Proteintech
Manufacturer Code:276101AP150UL
Catalog # 50-173-5716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FBXO11 Fusion Protein Ag24401 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F box only protein 11 F box protein 11 FBX11 FBXO11 PRMT9 UBR6 UG063H01 VIT 1 VIT1 Vitiligo associated protein 1; F-box only protein 11; Protein arginine N-methyltransferase 9; ubiquitin protein ligase E3 component n-recognin 6; VIT-1; Vitiligo-associated protein 1; vitiligo-associated protein VIT-1
Gene Aliases: FBX11; FBXO11; PRMT9; UBR6; UG063H01; VIT1
UniProt ID: (Human) Q86XK2
Entrez Gene ID: (Human) 80204
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.