Abnova
Manufacturer Code:PAB6454
Catalog # 89-132-415
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human FBXL10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of FBXL10.
For Research Use Only
Protein Aliases: [Histone-H3]-lysine-36 demethylase 1B; CXXC-type zinc finger protein 2; CXXC2; F-box and leucine-rich repeat protein 10; F-box protein FBL10; F-box/LRR-repeat protein 10; Fbl10; JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein; JHDM1B; JmjC domain-containing histone demethylation protein 1B; jumonji C domain-containing histone demethylase 1B; Jumonji domain-containing EMSY-interactor methyltransferase motif protein; KDM2B; lysine (K)-specific demethylase 2B; Lysine-specific demethylase 2B; PCCX2; Protein JEMMA; Protein-containing CXXC domain 2
Gene Aliases: CXXC2; FBL10; FBXL10; JHDM1B; KDM2B; NDY1; PCCX2
UniProt ID: (Human) Q8NHM5
Entrez Gene ID: (Human) 84678
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.