Invitrogen
Manufacturer Code:PA576444
Catalog # PIPA576444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human FBP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-fructose-1,6-bisphosphate 1-phosphohydrolase 2; FBPase 2; FBPase 2 D-fructose-16-bisphosphate 1-phosphohydrolase 2 Muscle FBPase FBP2; FBPase muscle; fructose bisphosphatase 2; Fructose-1,6-bisphosphatase isozyme 2; Muscle FBPase; RAE-30
Gene Aliases: Fbp-1; Fbp1; Fbp2; FBPase; Rae-30
UniProt ID: (Rat) Q9Z1N1, (Mouse) P70695
Entrez Gene ID: (Rat) 114508, (Mouse) 14120
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.