Invitrogen
Manufacturer Code:PA513689
Catalog # PIPA513689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 250-281 amino acids from the C-terminal region of human FBP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-fructose-1,6-bisphosphate 1-phosphohydrolase 2; FBPase 2; FBPase muscle; fructose-1,6-bisphosphatase 2; Fructose-1,6-bisphosphatase isozyme 2; Fructose-16-bisphosphatase isozyme 2 FBPase 2 D-fructose-16-bisphosphate 1-phosphohydrolase 2 FBP2; hexosediphosphatase; Muscle FBPase; muscle fructose-bisphosphatase; RAE-30
Gene Aliases: Fbp-1; Fbp1; FBP2; Rae-30
UniProt ID: (Human) O00757, (Mouse) P70695
Entrez Gene ID: (Human) 8789, (Mouse) 14120
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.