Invitrogen
Manufacturer Code:PA552943
Catalog # PIPA552943
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FBP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 2; FBPase 1; FBPase 2; Fructose-1,6-bisphosphatase 1; fructose-1,6-bisphosphatase 2; Fructose-1,6-bisphosphatase isozyme 2; Fructose-16-bisphosphatase isozyme 2 FBPase 2 D-fructose-16-bisphosphate 1-phosphohydrolase 2 FBP2 D-fructose-16-bisphosphate 1-phosphohydrolase 1 FBP1 FBPase 1 Fructose-16-bisphosphatase 1 growth-inhibiting protein 17 Liver FBPase; growth-inhibiting protein 17; hexosediphosphatase; Liver FBPase; Muscle FBPase; muscle fructose-bisphosphatase
Gene Aliases: FBP; FBP1; FBP2
UniProt ID: (Human) P09467, (Human) O00757
Entrez Gene ID: (Human) 2203, (Human) 8789
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.