Invitrogen
Manufacturer Code:MA535149
Catalog # PIMA535149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0664 |
Immunogen | A synthesized peptide derived from human FBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: BOS_9181 D-fructose-16-bisphosphate 1-phosphohydrolase 1 EC 3.1.3.11 Fbp FBP 1 FBP1 Fbp2 Fbp-2 Fbp3 FBPase 1 FBPase 3 Fdp Fructose-16-bisphosphatase 1 Fructose-16-bisphosphatase isozyme 3 Liver FBPase; D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; fructose-1,6- biphosphatase 1; Fructose-1,6-bisphosphatase 1; Fructose-1,6-bisphosphatase isozyme 3; Fructose-16- biphosphatase; growth-inhibiting protein 17; Liver FBPase
Gene Aliases: FBP; Fbp-2; FBP1; Fbp2; Fbp3; Fdp
UniProt ID: (Human) P09467, (Rat) P19112, (Mouse) Q9QXD6
Entrez Gene ID: (Human) 2203, (Rat) 24362, (Mouse) 14121
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.