Invitrogen
Manufacturer Code:PA530028
Catalog # PIPA530028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 51 and 338 of Human FBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-fructose-1,6-bisphosphate 1-phosphohydrolase 1; D-fructose-1,6-bisphosphate 1-phosphohydrolase 3; FBP D-fructose-16-bisphosphate 1-phosphohydrolase 1 FBPase 1 fructose-bisphosphatase 1 growth-inhibiting protein 17; FBPase 1; FBPase 3; FBPase brain isoform; FBPase liver; Fructose-1,6-bisphosphatase 1; Fructose-1,6-bisphosphatase isozyme 3; growth-inhibiting protein 17; Liver FBPase
Gene Aliases: FBP; Fbp-2; FBP1; Fbp2; Fbp3
UniProt ID: (Human) P09467, (Mouse) Q9QXD6
Entrez Gene ID: (Human) 2203, (Mouse) 14121
Molecular Function:
carbohydrate phosphatase
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.