R&D Systems
Manufacturer Code:FAB3897G100UG
Catalog # FAB3897G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 390917 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human FATP5 Met1-Leu690 Accession # Q9Y2P5 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: ACSB ACSBBACS ACSVL6 ACSVL6BAL BA-CoA ligase Bile acid-CoA ligase bile acyl-CoA synthetase Cholate--CoA ligase EC 6.2.1.7 FACVL3 FACVL3FATP-5 FATP5Solute carrier family 27 member 5 Fatty acid transport protein 5 Fatty-acid-coenzyme A ligase very long-chain 3 FLJ22987 SLC27A5 solute carrier family 27 (fatty acid transporter) member 5 VLACSR VLACSRVery long-chain acyl-CoA synthetase homolog 2 VLCSH2 VLCSH2Very long-chain acyl-CoA synthetase-related protein VLCS-H2VLACS-related; BA-CoA ligase; BACS; Bile acid-CoA ligase; Bile acyl-CoA synthetase; Cholate--CoA ligase; FATP-5; Fatty acid transport protein 5; Fatty-acid-coenzyme A ligase, very long-chain 3; solute carrier family 27 (fatty acid transporter), member 5; Solute carrier family 27 member 5; Very long-chain acyl-CoA synthetase homolog 2; Very long-chain acyl-CoA synthetase-related protein; VLACS-related; VLCS-H2
Gene Aliases: ACSB; ACSVL6; BACS; BAL; FACVL3; FATP-5; FATP5; SLC27A5; VLACSR; VLCS-H2; VLCSH2
UniProt ID: (Human) Q9Y2P5
Entrez Gene ID: (Human) 10998
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.