R&D Systems
Manufacturer Code:FAB3650G100UG
Catalog # FAB3650G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 342142 |
Immunogen | NS0 mouse myeloma cell line transfected with human FATP4 Met1-Leu643 Accession # Q6P1M0 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: ACSVL4; ACSVL4 ACSVL4Solute carrier family 27 member 4 EC 6.2.1 EC 6.2.1.- EC 6.2.1.7 FATP-4 FATP4Fatty acid transport protein 4 IPS long-chain fatty acid transport protein 4 SLC27A4 solute carrier family 27 (fatty acid transporter) member 4; Arachidonate--CoA ligase; FATP-4; Long-chain fatty acid transport protein 4; Long-chain-fatty-acid--CoA ligase; solute carrier family 27 (fatty acid transporter), member 4; Solute carrier family 27 member 4; Very long-chain acyl-CoA synthetase 4
Gene Aliases: ACSVL4; FATP4; IPS; SLC27A4
UniProt ID: (Human) Q6P1M0
Entrez Gene ID: (Human) 10999
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.