R&D Systems
Manufacturer Code:IC4659S100UG
Catalog # IC4659S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 466106 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human FATP2 Met1-Leu620 Accession # O14975 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: ACSVL1 ACSVL1FATP-2 EC 6.2.1 EC 6.2.1.- EC 6.2.1.3 EC 6.2.1.7 FACVL1 FACVL1very long-chain fatty-acid-coenzyme A ligase 1 FATP2THCA-CoA ligase Fatty acid transport protein 2 Fatty-acid-coenzyme A ligase very long-chain 1 hFACVL1 HsT17226 Long-chain-fatty-acid--CoA ligase SLC27A2 solute carrier family 27 (fatty acid transporter) member 2 Solute carrier family 27 member 2 THCA-CoA ligase very-long-chain acyl-CoA synthetase VLACS VLACSVery long-chain-fatty-acid-CoA ligase VLCS VLCSvery long-chain acyl-CoA synthetase; Arachidonate--CoA ligase; FATP-2; Fatty acid transport protein 2; Fatty-acid-coenzyme A ligase, very long-chain 1; Long-chain-fatty-acid--CoA ligase; Phytanate--CoA ligase; solute carrier family 27 (fatty acid transporter), member 2; Solute carrier family 27 member 2; THCA-CoA ligase; Very long-chain acyl-CoA synthetase; very long-chain fatty-acid-coenzyme A ligase 1; Very long-chain-fatty-acid-CoA ligase; very-long-chain acyl-CoA synthetase; VLACS
Gene Aliases: ACSVL1; FACVL1; FATP2; hFACVL1; HsT17226; SLC27A2; VLACS; VLCS
UniProt ID: (Human) O14975
Entrez Gene ID: (Human) 11001
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.