R&D Systems
Manufacturer Code:IC3304G100UG
Catalog # IC3304G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 308420 |
Immunogen | NS0 mouse myeloma cell line transfected with human FATP1 Met1-Leu646 Accession # Q6PCB7 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: ACSVL5FATP-1 EC 6.2.1 EC 6.2.1.- FATP1Solute carrier family 27 member 1 FATPlong-chain fatty acid transport protein 1 Fatty acid transport protein 1 FLJ00336 MGC71751 SLC27A1 solute carrier family 27 (fatty acid transporter) member 1; FATP-1; fatty acid transport protein 1; Long-chain fatty acid transport protein 1; solute carrier family 27 (fatty acid transporter), member 1; Solute carrier family 27 member 1
Gene Aliases: ACSVL5; FATP; FATP-1; FATP1; SLC27A1
UniProt ID: (Human) Q6PCB7
Entrez Gene ID: (Human) 376497
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.