Proteintech
Manufacturer Code:174641AP150UL
Catalog # 50-173-1587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FASTKD2 Fusion Protein Ag11314 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAST kinase domain-containing protein 2; FAST kinase domain-containing protein 2, mitochondrial; FAST kinase domains 2 FASTKD2 KIAA0971
Gene Aliases: 2810421I24Rik; FASTKD2; KIAA0971; RGD1307883
UniProt ID: (Human) Q9NYY8, (Mouse) Q922E6, (Rat) Q5M7V7
Entrez Gene ID: (Human) 22868, (Mouse) 75619, (Rat) 301463
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.