Abnova
Manufacturer Code:H00002194M03
Catalog # 89-022-059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S1 |
Immunogen | FASN (AAH07909 1 a.a. ~ 439 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant FASN.
For Research Use Only
Protein Aliases: 3-hydroxyacyl-[acyl-carrier-protein] dehydratase; 3-oxoacyl-[acyl-carrier-protein] reductase; 3-oxoacyl-[acyl-carrier-protein] synthase; [Acyl-carrier-protein] S-acetyltransferase; [Acyl-carrier-protein] S-malonyltransferase; Acyl-[acyl-carrier-protein] hydrolase; Enoyl-[acyl-carrier-protein] reductase; FAS; Fatty acid synthase; MGC14367; MGC15706; OA-519; SDR27X1; short chain dehydrogenase/reductase family 27X, member 1; Type I fatty acid synthase
Gene Aliases: FAS; FASN; OA-519; SDR27X1
UniProt ID: (Human) P49327
Entrez Gene ID: (Human) 2194
Molecular Function: acyltransferase dehydrogenase esterase hydrolase ligase methyltransferase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.