Novus Biologicals
Manufacturer Code:NBP190435
Catalog # NBP190435
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:FLGKVLMEKLFRTSPDLKVIYILVRPKAGQTLQQRVFQILDSKLFEKVKEVCPNVHEKIRAIYADLNQNDFAISKEDMQELLSCTNIIFHCAATVRFDD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 81; fatty acyl CoA reductase 2; fatty acyl CoA reductase 2 FLJ10462 member 2 MLSTD1; Fatty acyl-CoA reductase 2; Male sterility domain-containing protein 1; short chain dehydrogenase/reductase family 10E, member 2
Gene Aliases: FAR2; HEL-S-81; MLSTD1; SDR10E2
UniProt ID: (Human) Q96K12
Entrez Gene ID: (Human) 55711
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.