Abnova
Manufacturer Code:PAB20854
Catalog # 89-122-351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human FAR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant FAR2.
For Research Use Only
Protein Aliases: epididymis secretory protein Li 81; fatty acyl CoA reductase 2; Fatty acyl-CoA reductase 2; FLJ10462; Male sterility domain-containing protein 1; MLSTD1; SDR10E2; short chain dehydrogenase/reductase family 10E, member 2
Gene Aliases: FAR2; HEL-S-81; MLSTD1; SDR10E2
UniProt ID: (Human) Q96K12
Entrez Gene ID: (Human) 55711
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.