Invitrogen
Manufacturer Code:PA567238
Catalog # PIPA567238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human FAPP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAPP-2; PH domain-containing family A member 8; Phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8; Pleckstrin homology domain-containing family A member 8; PLEKHA8 FAPP2 phosphoinositol 4-phosphate adaptor protein-2 MGC3358 pleckstrin homology domain containing family A (phosphoinositide binding specific) member 8; Serologically defined breast cancer antigen NY-BR-86
Gene Aliases: FAPP2; PLEKHA8
UniProt ID: (Human) Q96JA3
Entrez Gene ID: (Human) 84725
Molecular Function: nucleic acid binding transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.