Invitrogen
Manufacturer Code:PA5113305
Catalog # PIPA5113305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Pleckstrin homology domain-containing family A member 8 protein (1-300AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6330400G01 AA517676 BC052360 Fapp2 FAPP-2 hFAPP2 MGC3358 PH domain-containing family A member 8 Phosphatidylinositol-four-phosphate adapter protein 2 phosphoinositol 4-phosphate adapter protein 2 pleckstrin homology domain containing A8 pleckstrin homology domain containing family A (phosphoinositide binding specific) member 8 Pleckstrin homology domain-containing family A member 8 Plekha8 RGD1563940 Serologically defined breast cancer antigen NY-BR-86; FAPP-2; PH domain-containing family A member 8; Phosphatidylinositol-four-phosphate adapter protein 2; phosphoinositol 4-phosphate adapter protein 2; pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8; Pleckstrin homology domain-containing family A member 8; Serologically defined breast cancer antigen NY-BR-86
Gene Aliases: 6330400G01; AA517676; BC052360; Fapp-2; FAPP2; PLEKHA8
UniProt ID: (Human) Q96JA3, (Mouse) Q80W71
Entrez Gene ID: (Human) 84725, (Mouse) 231999
Molecular Function:
nucleic acid binding
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.