Novus Biologicals
Manufacturer Code:NB100182
Catalog # NB100182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This FANCD2 Antibody was developed against human FANCD2 fusion protein (N-terminal fragment). [Swiss-Prot #Q9BXW9] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp762A223 FA4 FACD FAD FAD2 FA-D2 FADFAD2 FANCD Fanconi anemia complementation group D2 Fanconi anemia group D2 protein Fanconi anemia complementation group D2 FLJ23826 FPN1 HFE4 IREG1 Protein FACD2 SLC11A3; Fanconi anemia group D2 protein; Fanconi anemia, complementation group D2; Protein FACD2
Gene Aliases: FA-D2; FA4; FACD; FAD; FAD2; FANCD; FANCD2
UniProt ID: (Human) Q9BXW9
Entrez Gene ID: (Human) 2177
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.