Invitrogen
Manufacturer Code:PA546445
Catalog # PIPA546445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human FAM86A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eEF2-KMT; eEF2-lysine methyltransferase; family with sequence similarity 86, member A; protein FAM86A; Protein-lysine N-methyltransferase EEF2KMT; putative protein N-methyltransferase FAM86A
Gene Aliases: eEF2-KMT; EEF2KMT; FAM86A; SB153
UniProt ID: (Human) Q96G04
Entrez Gene ID: (Human) 196483
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.