Novus Biologicals
Manufacturer Code:NBP232219
Catalog # NBP232219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen this antibody was made to maps to a region between residue 1129 to 1179 of human Family with Sequence Similarity 83 Member H using the numbering given in entry NP_940890.3 (GeneID 286077). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAM83H variant 1; FAM83H variant 2; family with sequence similarity 83 member H protein FAM83H; family with sequence similarity 83, member H; Protein FAM83H
Gene Aliases: AI3; FAM83H
UniProt ID: (Human) Q6ZRV2
Entrez Gene ID: (Human) 286077
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.