Proteintech
Manufacturer Code:212031AP150UL
Catalog # 50-173-3316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FAM72 Fusion Protein Ag15566 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dpr4-related protein; FAM72 FAM72A FAM72B FAM72C FAM72D Protein FAM72A; family with sequence similarity 72, member A; Latent membrane protein 1-induced protein; LMP1-induced protein; Protein FAM72A
Gene Aliases: 2700049P18Rik; FAM72A; LMPIP; P17; UGENE
UniProt ID: (Human) Q5TYM5, (Rat) A1KXW8, (Mouse) Q8BFZ8
Entrez Gene ID: (Human) 729533, (Rat) 681249, (Mouse) 108900
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.