Proteintech
Manufacturer Code:207601AP150UL
Catalog # 50-173-3067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FAM57B Fusion Protein Ag14707 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase; FAM57B FP1188 Protein FAM57B; family with sequence similarity 57, member B; Protein FAM57B; TLC domain-containing protein 3B
Gene Aliases: 1500016O10Rik; A330104J06Rik; AI413816; AW060769; FAM57B; FP1188; RGD1308215; TLCD3B
UniProt ID: (Human) Q71RH2, (Mouse) Q7TNV1
Entrez Gene ID: (Human) 83723, (Mouse) 68952, (Rat) 293493
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.