Proteintech
Manufacturer Code:244191AP150UL
Catalog # 50-173-4347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FAM48A Fusion Protein Ag19875 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C13 C13orf19 FAM48A FP757 p38 interacting protein P38IP Protein FAM48A; family with sequence similarity 48, member A; p38-interacting protein; p38IP; protein FAM48A; suppressor of Ty 20 homolog; transcription factor (p38 interacting protein); Transcription factor SPT20 homolog; tumor rejection antigen
Gene Aliases: C13; C13orf19; FAM48A; FP757; P38IP; SPT20; SUPT20H
UniProt ID: (Human) Q8NEM7
Entrez Gene ID: (Human) 55578
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.