Novus Biologicals
Manufacturer Code:NBP243534
Catalog # NBP243534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human FAM21C. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Em:AC012044.3 family with sequence similarity 21 member C KIAA0592FAM21A VPEFVaccinia virus penetration factor WASH complex subunit FAM21C; family with sequence similarity 21, member C; Vaccinia virus penetration factor; VPEF; WASH complex subunit 2C
Gene Aliases: FAM21A; FAM21C; KIAA0592; VPEF; WASHC2C
UniProt ID: (Human) Q9Y4E1
Entrez Gene ID: (Human) 253725
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.