Invitrogen
Manufacturer Code:PA569890
Catalog # PIPA569890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human FAM213B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: family with sequence similarity 213 member B; family with sequence similarity 213, member B; Peroxiredoxin-like 2B; Prostamide/PG F synthase; prostamide/PGF synthase; Prostamide/prostaglandin F synthase; Protein FAM213B; prxl2
Gene Aliases: 2810405K02Rik; AI836168; C16H1orf93; C1orf93; C5H1orf93; FAM213B; PM/PGFS; PRXL2B; RGD1308251; sb:cb782; selu3; wu:fc33a02; zgc:85644
UniProt ID: (Human) B9DI92, (Mouse) Q9DB60, (Rat) D3ZVR7, (Bovine) Q58CY6, (Zebrafish) Q6NV24
Entrez Gene ID: (Human) 127281, (Mouse) 66469, (Rat) 362676, (Horse) 100054405, (Dog) 489616, (Bovine) 617001, (Guinea pig) 100733270, (Zebrafish) 406605
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.