Novus Biologicals
Manufacturer Code:NBP216415
Catalog # NBP216415
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human DMP4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dentin matrix protein 4; dentin matrix protein 4 DKFZp547C074 DMP4 DMP-4 family with sequence similarity 20 member C RNS; DMP-4; Extracellular serine/threonine protein kinase FAM20C; family with sequence similarity 20, member C; GEF-CK; Golgi casein kinase; Golgi-enriched fraction casein kinase
Gene Aliases: DMP-4; DMP4; FAM20C; GEF-CK; RNS
UniProt ID: (Human) Q9NPT2
Entrez Gene ID: (Human) 56975
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.