Invitrogen
Manufacturer Code:PA570352
Catalog # PIPA570352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human FAM170B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acrosome-related protein; family with sequence similarity 170, member B; Protein FAM170B; putative protein FAM170B; rCG42349-like
Gene Aliases: 4922501K12Rik; AI449756; C10orf73; FAM170B
UniProt ID: (Human) A6NMN3, (Mouse) E9PXT9
Entrez Gene ID: (Rabbit) 100358665, (Dog) 102153845, (Human) 170370, (Horse) 100629985, (Mouse) 105511, (Guinea pig) 100720326, (Rat) 100362169
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.