Invitrogen
Manufacturer Code:PA555591
Catalog # PIPA555591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human FAM150A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALK and LTK ligand 1; AUG-beta; Augmentor beta; family with sequence similarity 150, member A; Protein FAM150A; RPLK9433
Gene Aliases: ALKAL1; FAM150A; UNQ9433; UNQ9433/PRO34745
UniProt ID: (Human) Q6UXT8
Entrez Gene ID: (Human) 389658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.