Abnova
Manufacturer Code:PAB22025
Catalog # 89-123-733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human FAM150A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant FAM150A.
For Research Use Only
Protein Aliases: ALK and LTK ligand 1; AUG-beta; Augmentor beta; family with sequence similarity 150, member A; MGC163513; MGC163515; Protein FAM150A; RPLK9433; UNQ9433
Gene Aliases: ALKAL1; FAM150A; UNQ9433; UNQ9433/PRO34745
UniProt ID: (Human) Q6UXT8
Entrez Gene ID: (Human) 389658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.