Invitrogen
Manufacturer Code:PA589911
Catalog # PIPA589911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-200 of human FAM13A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAM13A1_v2 protein; family with sequence similarity 13, member A; family with sequence similarity 13, member A1; Precm1 protein; Protein FAM13A
Gene Aliases: AI462012; ARHGAP48; D430015B01Rik; FAM13A; FAM13A1; KIAA0914; Precm1; RGD1309807
UniProt ID: (Human) O94988, (Mouse) Q8BGI4
Entrez Gene ID: (Human) 10144, (Mouse) 58909, (Rat) 362378
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.