Novus Biologicals
Manufacturer Code:NBP2847040.1ML
Catalog # NB032632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human FAM13A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARHGAP48 FAM13A1 FAM13A1_v2 protein family with sequence similarity 13 member A KIAA0914 member A1 MGC105131; FAM13A1_v2 protein; family with sequence similarity 13, member A; family with sequence similarity 13, member A1; Protein FAM13A
Gene Aliases: ARHGAP48; FAM13A; FAM13A1; KIAA0914
UniProt ID: (Human) O94988
Entrez Gene ID: (Human) 10144
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.