Invitrogen
Manufacturer Code:PA542647
Catalog # PIPA542647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human FAM134B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: family with sequence similarity 134 member B; family with sequence similarity 134, member B; protein FAM134B; Protein FAM134B-like protein; Reticulophagy receptor 1; reticulophagy receptor FAM134B; Reticulophagy regulator 1
Gene Aliases: 1810015C04Rik; AU015349; FAM134B; JK-1; JK1; RETREG1
UniProt ID: (Human) Q9H6L5, (Rat) Q5FVM3, (Mouse) Q8VE91, (Bovine) Q5E9K8
Entrez Gene ID: (Human) 54463, (Rat) 619558, (Mouse) 66270, (Dog) 479382, (Rabbit) 100343570, (Horse) 100147549, (Bovine) 540068, (Guinea pig) 100720866
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.