Invitrogen
Manufacturer Code:PA567448
Catalog # PIPA567448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human FAM132B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1QTNF15 CTRP15 ERFE; Complement C1q tumor necrosis factor-related protein 15; Erythroferrone; family with sequence similarity 132 member B; family with sequence similarity 132, member B; Myonectin; protein FAM132B
Gene Aliases: C1QTNF15; CTRP15; ERFE; FAM132B
UniProt ID: (Human) Q4G0M1
Entrez Gene ID: (Human) 151176
Molecular Function: extracellular matrix protein extracellular matrix structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.