Abnova
Manufacturer Code:H00010876M01
Catalog # 89-106-419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D9 |
Immunogen | FAM12A (NP_006674.21 a.a. &bsim 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant FAM12A.
For Research Use Only
Protein Aliases: EP3A; epididymal secretory protein E3 alpha; Epididymal secretory protein E3-alpha; epididymis-specific 3 alpha; family with sequence similarity 12, member A; HE3-ALPHA; HE3A; HE3ALPHA; human epididymis-specific 3 alpha; Human epididymis-specific protein 3-alpha; MGC119614; MGC119615; ribonuclease A M1
Gene Aliases: EDDM3A; EP3A; FAM12A; HE3-ALPHA; HE3A; HE3ALPHA; RAM1
UniProt ID: (Human) Q14507
Entrez Gene ID: (Human) 10876
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.