Novus Biologicals
Manufacturer Code:NBP213945
Catalog # NBP213945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: KEALKGKSFHMFIYSLWFKIQRACINEKGSDRYRNAYVWAPGALKVLECH WEKYNNRYTESRSFS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EP3A epididymal protein 3A epididymal secretory protein E3-alpha epididymis-specific 3 alpha FAM12AMGC119615 family with sequence similarity 12 member A HE3Aepididymal secretory protein E3 alpha HE3-alpha HE3ALPHA human epididymis-specific 3 alpha Human epididymis-specific protein 3-alpha MGC119614; epididymal secretory protein E3 alpha; Epididymal secretory protein E3-alpha; epididymis-specific 3 alpha; family with sequence similarity 12, member A; HE3-alpha; human epididymis-specific 3 alpha; Human epididymis-specific protein 3-alpha; ribonuclease A M1
Gene Aliases: EDDM3A; EP3A; FAM12A; HE3-ALPHA; HE3A; HE3ALPHA; RAM1
UniProt ID: (Human) Q14507
Entrez Gene ID: (Human) 10876
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.