Invitrogen
Manufacturer Code:PA569874
Catalog # PIPA569874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human METTL21A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: family with sequence similarity 119, member A; heat shock protein 70kDa lysine (K) methyltransferase; Hepatocellular carcinoma-associated antigen 557b; HSPA lysine methyltransferase; HSPA-KMT; Methyltransferase-like protein 21A; Protein N-lysine methyltransferase METTL21A; protein N-lysine methyltransferase METTL21A-like; protein-lysine methyltransferase METTL21A
Gene Aliases: 2310038H17Rik; AI464204; FAM119A; HCA557B; HSPA-KMT; METTL21A
UniProt ID: (Bovine) A4FV42, (Human) Q8WXB1, (Mouse) Q9CQL0
Entrez Gene ID: (Rat) 102553119, (Dog) 607933, (Bovine) 615773, (Human) 151194, (Horse) 100068972, (Rabbit) 100352407, (Guinea pig) 100727792, (Mouse) 67099
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.