Novus Biologicals
Manufacturer Code:NBP216410
Catalog # NBP216410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human FAM115C. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: family with sequence similarity 115 member C member A; family with sequence similarity 115, member C; family with sequence similarity 139, member A; protein FAM115C; TRP channel-associated factor 2; TRPM8 channel-associated factor 2
Gene Aliases: FAM115C; FAM139A; TCAF2
UniProt ID: (Human) A6NFQ2
Entrez Gene ID: (Human) 285966
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.