Novus Biologicals
Manufacturer Code:NBP189407
Catalog # NBP189407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SATVATVGQGISNVIEKAETSLGIPSPSEISTEVKYVAGETNAKENENSSPVAGAFGVFSTISTAVQSTGKSVISG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C5orf3 chromosome 5 open reading frame 3 family with sequence similarity 114 member A2133K02 hypothetical protein LOC10827; Protein FAM114A2; testis tissue sperm-binding protein Li 81P
Gene Aliases: 133K02; C5orf3; FAM114A2
UniProt ID: (Human) Q9NRY5
Entrez Gene ID: (Human) 10827
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.