Novus Biologicals
Manufacturer Code:NBP157739
Catalog # NBP157739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to FAM109A (family with sequence similarity 109 member A) The peptide sequence was selected from the N terminal of FAM109A)(50ug). Peptide sequence HRRWFVLRGNMLFYFEDAASREPVGVIILEGCTVELVEAAEEFAFAVRFA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 27 kDa inositol polyphosphate phosphatase-interacting protein A; 27 kDa inositol polyphosphate phosphatase-interacting protein A family with sequence similarity 109 member A FLJ32356 IPIP27A protein FAM109A SES1; family with sequence similarity 109, member A; IPIP27A; PH domain-containing endocytic trafficking adaptor 1; protein FAM109A; Ses1; Sesquipedalian-1
Gene Aliases: FAM109A; IPIP27A; PHETA1; SES1
UniProt ID: (Human) Q8N4B1
Entrez Gene ID: (Human) 144717
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.