Invitrogen
Manufacturer Code:PA546020
Catalog # PIPA546020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human FAM108A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: abhydrolase domain containing 17A; Abhydrolase domain-containing protein 17A; abhydrolase domain-containing protein FAM108A; abhydrolase domain-containing protein FAM108A1; Alpha/beta hydrolase domain-containing protein 17A; family with sequence similarity 108, member A; family with sequence similarity 108, member A1; mFLJ00358 protein; protein ABHD17A
Gene Aliases: 1700013O15Rik; ABHD17A; BC005632; C19orf27; D10Bwg1364e; FAM108A; FAM108A1; RGD1359682; upf0227
UniProt ID: (Human) Q96GS6, (Bovine) Q2HJ19, (Mouse) Q99JW1, (Rat) Q5XIJ5
Entrez Gene ID: (Dog) 485076, (Human) 81926, (Horse) 100146322, (Bovine) 614425, (Guinea pig) 100733633, (Mouse) 216169, (Rat) 299617
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.