Invitrogen
Manufacturer Code:PA570893
Catalog # PIPA570893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of rat FAM107B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: axoneme-associated protein mst101(2)-like; family with sequence similarity 107, member B; novel DUF1151 domain containing protein; Protein FAM107B
Gene Aliases: 3110001A13Rik; AA589595; AA959793; AI132312; C10orf45; FAM107B; wu:fc52f02; zgc:56216
UniProt ID: (Human) Q9H098, (Bovine) Q2KI00, (Rat) Q5U4F3, (Mouse) Q3TGF2
Entrez Gene ID: (Horse) 100056470, (Rabbit) 100341804, (Dog) 607279, (Human) 83641, (Bovine) 535023, (Guinea pig) 100727938, (Rat) 498796, (Mouse) 66540, (Zebrafish) 386641
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.