Novus Biologicals
Manufacturer Code:NBP287398
Catalog # NB035020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Rat FAM107B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C10orf45 chromosome 10 open reading frame 45 family with sequence similarity 107 member B FLJ45505 hypothetical protein LOC83641 MGC11034 MGC90261; family with sequence similarity 107, member B; Protein FAM107B
Gene Aliases: C10orf45; FAM107B
UniProt ID: (Human) Q9H098
Entrez Gene ID: (Human) 83641
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.