Invitrogen
Manufacturer Code:PA543349
Catalog # PIPA543349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human FAM105A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: family with sequence similarity 105 member A; family with sequence similarity 105, member A; inactive ubiquitin thioesterase FAM105A; Inactive ubiquitin thioesterase OTULINL; protein FAM105A; Protein FAM105A-like protein
Gene Aliases: 9830126M18; FAM105A; NET20; OTULINL
UniProt ID: (Human) Q9NUU6, (Mouse) Q3TVP5, (Rat) Q3B7D8
Entrez Gene ID: (Human) 54491, (Mouse) 223433, (Rat) 310190, (Dog) 606782, (Bovine) 534389, (Horse) 100070247, (Rabbit) 100342296
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.