Novus Biologicals
Manufacturer Code:NBP213974
Catalog # NBP213974
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: RRRNGSKEGNHHSTQPKRNKRNPIFQDSQDTEFSWSDNERSSSRINIPER ASGPEGNLNQIVTEPDANFPQFLHEGLSKPVYVINWFMSFGPEIKLNTSQ QG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome X open reading frame 44 CXorf44 family with sequence similarity 104 member B FLJ18375 FLJ20434 hypothetical protein LOC90736; family with sequence similarity 104, member B; Protein FAM104B
Gene Aliases: CXorf44; FAM104B
UniProt ID: (Human) Q5XKR9
Entrez Gene ID: (Human) 90736
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.