Invitrogen
Manufacturer Code:PA570892
Catalog # PIPA570892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the Middle region of human FAM102A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C230093N12Rik; Early estrogen-induced gene 1 protein; FAM102A C9orf132 EEIG1; family with sequence similarity 102, member A; Protein FAM102A; sym-3 homolog A
Gene Aliases: bA203J24.7; C9orf132; EEIG1; FAM102A; SYM-3A
UniProt ID: (Human) Q5T9C2
Entrez Gene ID: (Human) 399665
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.