Invitrogen
Manufacturer Code:PA570891
Catalog # PIPA570891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human FAM101B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: family with sequence similarity 101, member B; filamin-interacting protein FAM101B; protein FAM101B; Refilin-B; RefilinB; Regulator of filamin protein B
Gene Aliases: 1500005K14Rik; cfm; CFM1; FAM101B; RefilinB; RFLNB; RGD1359691
UniProt ID: (Human) Q8N5W9, (Rat) Q6AXS9, (Mouse) Q5SVD0
Entrez Gene ID: (Human) 359845, (Rat) 287534, (Mouse) 76566, (Dog) 611214, (Pig) 100524216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.