Proteintech
Manufacturer Code:211131AP150UL
Catalog # 50-173-3263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | FAM101A Fusion Protein Ag15314 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAM101A Protein FAM101A; family with sequence similarity 101, member A; filamin-interacting protein FAM101A; protein FAM101A; Refilin-A; RefilinA; Regulator of filamin protein A
Gene Aliases: 3110032G18Rik; cfm; CFM2; FAM101A; RFLNA
UniProt ID: (Human) Q6ZTI6, (Mouse) Q7TS73
Entrez Gene ID: (Human) 144347, (Mouse) 73121, (Rat) 689711
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.