R&D Systems
Manufacturer Code:FAB9494N100UG
Catalog # FAB9494N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 992338 |
Immunogen | Human embryonic kidney cell line HEK293-derived recombinant human Fc mu R/FAIM3 Arg18-Gly251 Accession # O60667 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: FAIM3 Fas apoptotic inhibitory molecule 3 Fc mu receptor FCMR immunoglobulin mu Fc receptor Regulator of Fas-induced apoptosis Toso Toso TOSOIgM Fc receptor; Fas apoptotic inhibitory molecule 3; Fc mu receptor; IgM Fc fragment receptor; IgM Fc receptor; immunoglobulin mu Fc receptor; Regulator of Fas-induced apoptosis Toso
Gene Aliases: FAIM3; FCMR; TOSO
UniProt ID: (Human) O60667
Entrez Gene ID: (Human) 9214
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.