Novus Biologicals
Manufacturer Code:NBP186924
Catalog # NBP186924
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RGKTQKVTLNVHSEYEPSWEEQPMPETPKWFHLPYLFQMPAYASSSKFVTRVTTPAQRGKVPPVHHSSPTTQITHRPRVSRASSVAGDKPRTFLPSTTASKISALEGLLKPQTPSYNHHTRLHRQRALDYGSQSGRE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fas apoptotic inhibitory molecule 3; Fas apoptotic inhibitory molecule 3 Fc mu receptor FCMR immunoglobulin mu Fc receptor Regulator of Fas-induced apoptosis Toso TOSOIgM Fc receptor; Fc mu receptor; IgM Fc fragment receptor; IgM Fc receptor; immunoglobulin mu Fc receptor; Regulator of Fas-induced apoptosis Toso
Gene Aliases: FAIM3; FCMR; TOSO
UniProt ID: (Human) O60667
Entrez Gene ID: (Human) 9214
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.