Abnova
Manufacturer Code:PAB12849
Catalog # 89-116-129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 13 amino acids of human FAIM3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of FAIM3.
For Research Use Only
Protein Aliases: Fas apoptotic inhibitory molecule 3; Fc mu receptor; IgM Fc fragment receptor; IgM Fc receptor; immunoglobulin mu Fc receptor; Regulator of Fas-induced apoptosis Toso; TOSO
Gene Aliases: FAIM3; FCMR; TOSO
UniProt ID: (Human) O60667
Entrez Gene ID: (Human) 9214
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.